1,3-Dimethoxybenzene 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Resorcinol dimethyl ether

15523-1000 15523-5000
TS15523-1000EA 37 USD
TS15523-1000 TS15523-5000
1,3-Dimethoxybenzene 99%
1,3-Dimethoxybenzene
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 85…87 °C (7 mmHg)
Melting Pt: –52 °C
Density: 1.05 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00008384
CAS Number: 151-10-0
EINECS: 205-783-4

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5230 to 1.5250 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR