2,3-Dihydroxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

15517-0050 15517-0250
TS15517-0050EA 56 USD
TS15517-0050 TS15517-0250
2,3-Dihydroxybenzoic acid 99%
2,3-Dihydroxybenzoic acid
Formula: C₇H₆O₄
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 204…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00002446
CAS Number: 303-38-8
EINECS: 206-139-5

Order Now

Specification Test Results

Appearance Beige to pink to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 204 to 208 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR