2,5-Dibromoaniline 97%

Supplier: Thermo Scientific Chemicals
Danger

15503-0050 15503-0250
TS15503-0050EA 49 USD
TS15503-0050 TS15503-0250
2,5-Dibromoaniline 97%
2,5-Dibromoaniline
Formula: C₆H₅Br₂N
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 51…56 °C
Flash Pt: 109 °C
MDL Number: MFCD00007636
CAS Number: 3638-73-1
EINECS: 222-865-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to dark grey Crystalline powder
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR