4-Chlorothiophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Chloro-4-mercaptobenzene, 4-Chlorobenzene-1-thiol, 4-Chlorobenzenethiol, p-Chlorothiophenol

15476-0050 15476-1000 15476-5000
TS15476-0050EA 29 USD
TS15476-0050 TS15476-1000 TS15476-5000
4-Chlorothiophenol 98%
4-Chlorothiophenol
Formula: C₆H₅ClS
MW: 144.62 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 49…53 °C
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00004847
CAS Number: 106-54-7
EINECS: 203-408-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
Melting point 49 to 53 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR