2-Chloro-p-toluidine 98%

Supplier: Thermo Scientific Chemicals
Danger

15467-0050 15467-0250
TS15467-0050EA 19 USD
TS15467-0050 TS15467-0250
2-Chloro-p-toluidine 98%
2-Chloro-p-toluidine
Formula: C₇H₈ClN
Boiling Pt: 223…225 °C (760 mmHg)
Melting Pt: 7 °C
Density: 1.151 g/cm³
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD00007666
CAS Number: 615-65-6
EINECS: 210-440-7
UN: 3429
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5740 to 1.5760 (20°C, 589 nm)
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR