3-tert-Butylphenol 99+%

Supplier: Thermo Scientific Chemicals
Danger

15438-0010 15438-0250 15438-1000
TS15438-0010EA 26 USD
TS15438-0010 TS15438-0250 TS15438-1000
3-tert-Butylphenol 99+%
3-tert-Butylphenol
Formula: C₁₀H₁₄O
Boiling Pt: 125…130 °C (20 mmHg)
Melting Pt: 40…43 °C
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00002300
CAS Number: 585-34-2
EINECS: 209-553-4
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 40 to 43 °C
GC ≥99.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR