1,4-Butanediol diglycidyl ether 60%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Araldite RD-2, 1,4-Bis(oxiran-2-ylmethoxy)butane
15431-0025
15431-5000
TS15431-0025EA
1033
USD
TS15431-0025
TS15431-5000
1,4-Butanediol diglycidyl ether 60%, Technical Grade
1,4-Butanediol diglycidyl ether
Formula:
C₁₀H₁₈O₄ MW: 202.25 g/mol Boiling Pt: 266 °C (760 mmHg) Density: 1.1 g/cm³ Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005146 CAS Number: 2425-79-8 EINECS: 219-371-7 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4590 to 1.4650 (20°C, 589 nm) |
GC | ≥55 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...