2-Fluorobenzoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: o-Fluorobenzoyl chloride

15309-0250 15309-1000
TS15309-0250EA 45 USD
TS15309-0250 TS15309-1000
2-Fluorobenzoyl chloride 97%
2-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 90…92 °C (15 mmHg)
Melting Pt: 4 °C
Density: 1.32 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000656
CAS Number: 393-52-2
EINECS: 206-887-2
UN: 1760
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.5355 to 1.5375 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR