4-Nitrobenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals
Warning

15302-0050 15302-0250
TS15302-0050EA 23 USD
TS15302-0050 TS15302-0250
4-Nitrobenzenesulfonamide 97%
4-Nitrobenzenesulphonamide
Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Melting Pt: 178…182 °C
MDL Number: MFCD00007937
CAS Number: 6325-93-5
EINECS: 228-691-6

Order Now

Specification Test Results

Appearance White to beige to tan Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 175 to 181 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR