4-Fluorobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Fluorobenzoyl chloride

15295-0250 15295-1000
TS15295-0250EA 53 USD
TS15295-0250 TS15295-1000
4-Fluorobenzoyl chloride 98%
4-Fluorobenzoyl chloride
Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 82 °C (20 mmHg)
Melting Pt: 9 °C
Density: 1.34 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000684
CAS Number: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5299 to 1.5319 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR