2,4,6-Triisopropylbenzenesulphonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,4,6-Triisopropylbenzene-1-sulphonyl chloride
15279-0250
15279-1000
TS15279-0250EA
74
USD
TS15279-0250
TS15279-1000
2,4,6-Triisopropylbenzenesulphonyl chloride 97%
2,4,6-Triisopropylbenzenesulphonyl chloride
Formula:
C₁₅H₂₃ClO₂S Melting Pt: 93…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007433 CAS Number: 6553-96-4 EINECS: 229-479-6 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to light beige Crystalline solid |
Infrared spectrum | Conforms |
Melting point | 93 to 97 °C |
Titration Argentometric | ≥96.0 % |
Gel strength | ≥700 g/cm² (1.5 %) |
Gel point | 32 to 38 °C (1.5 %) |
Ash | ≤5 % |
pH | 6.0 to 7.5 (10 % suspension) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...