2-Cyanophenylacetonitrile 80%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Cyanomethyl)benzonitrile, α-Cyano-o-tolunitrile, Homophthalonitrile

15257-0250 15257-1000
TS15257-0250EA 51 USD
TS15257-0250 TS15257-1000
2-Cyanophenylacetonitrile 80%
2-Cyanophenylacetonitrile
Formula: C₉H₆N₂
MW: 142.16 g/mol
Melting Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00001895
CAS Number: 3759-28-2
EINECS: 223-167-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Beige to brown Powder and/or chunks
Infrared spectrum Conforms
GC ≥75 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR