trans-Anethole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (E)-1-Methoxy-4-(prop-1-en-1-yl)benzene

15253-0050 15253-1000 15253-5000
TS15253-0050EA 21 USD
TS15253-0050 TS15253-1000 TS15253-5000
trans-Anethole 99%
trans-Anethole
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 23 °C
Density: 0.9875 g/cm³
Flash Pt: 90 °C
MDL Number: MFCD00009284
CAS Number: 4180-23-8
EINECS: 224-052-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5590 to 1.5630 (20°C, 589 nm)
Density (20/4°C) 0.986 to 0.989
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR