m-Anisic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Anisic acid, 3-Methoxybenzoic acid

15247-0250 15247-1000 15247-5000
TS15247-0250EA 45 USD
TS15247-0250 TS15247-1000 TS15247-5000
m-Anisic acid 98%
m-Anisic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 170…172 °C (10 mmHg)
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00002499
CAS Number: 586-38-9
EINECS: 209-574-9

Order Now

Specification Test Results

Appearance Off-white to yellow to light beige Powder
Infrared spectrum Conforms
Melting point 103 to 108 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR