p-Anisic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methoxybenzoic acid, 4-Anisic acid

15237-0010 15237-0051 15237-1000 15237-2500
TS15237-0010EA 269 USD
TS15237-0010 TS15237-0051 TS15237-1000 TS15237-2500
p-Anisic acid 98%
p-Anisic acid
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 275 °C (760 mmHg)
Melting Pt: 181…186 °C
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002542
CAS Number: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669

Order Now

Specification Test Results

Appearance White to beige to light grey Powder
Infrared spectrum Conforms
Melting point 181 to 186 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR