2-Bromo-m-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-1,3-dimethylbenzene, 2,6-Dimethylbromobenzene
15203-0050
15203-0250
15203-1000
TS15203-0050EA
36
USD
TS15203-0050
TS15203-0250
TS15203-1000
2-Bromo-m-xylene 98%
2-Bromo-m-xylene
Formula:
C₈H₉Br MW: 185.06 g/mol Boiling Pt: 206 °C (760 mmHg) Melting Pt: –10 °C Density: 1.38 g/cm³ Flash Pt: 73 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000075 CAS Number: 576-22-7 EINECS: 209-397-7 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5540 to 1.5560 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | Approx. 100 ppm BHT |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...