2,2'-Dihydroxybiphenyl 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2'-Biphenol, 2,2-Biphenol, Biphenyl-2,2'-diol
15199-1000
15199-5000
TS15199-1000EA
50
USD
TS15199-1000
TS15199-5000
2,2'-Dihydroxybiphenyl 99%
2,2'-Dihydroxybiphenyl
Formula:
C₁₂H₁₀O₂ Boiling Pt: 315 °C (1013 hPa) Melting Pt: 108…111 °C Density: 0.83 g/cm³ Flash Pt: 154 °C |
MDL Number:
MFCD00002210 CAS Number: 1806-29-7 EINECS: 217-303-0 |
Specification Test Results
Appearance | Beige to light grey Powder |
Infrared spectrum | Conforms |
Melting point | 108 to 111 °C |
Loss on drying | ≤0.5 % |
GC | ≥98.5 % |
Iron (Fe) | ≤15 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...