4-Methoxybenzyl mercaptan 95%

Supplier: Thermo Scientific Chemicals
Danger

15095-0250 15095-1000
TS15095-0250EA 172 USD
TS15095-0250 TS15095-1000
4-Methoxybenzyl mercaptan 95%
4-Methoxybenzyl mercaptan
Formula: C₈H₁₀OS
Boiling Pt: 90…95 °C (.5 mmHg)
Density: 1.107 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004871
CAS Number: 6258-60-2
EINECS: 228-393-6
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5720 to 1.5780 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR