2,5-Di-tert-butylhydroquinone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: BHQ

15084-0010 15084-0025
TS15084-0010EA 118 USD
TS15084-0010 TS15084-0025
2,5-Di-tert-butylhydroquinone 99%
2,5-Di-tert-butylhydroquinone
Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Melting Pt: 212…218 °C
Flash Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00008825
CAS Number: 88-58-4
EINECS: 201-841-8

Order Now

Specification Test Results

Appearance Light beige to pink-beige Crystalline powder
Infrared spectrum Conforms
Melting point 212 to 218 °C
HPLC ≥98.5 %
Specific optical rotation +131° to +137° (20 °C, 589 nm) (c=1,MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR