4-Acetamidobenzenesulphonyl chloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Acetylsulphanilyl chloride

15072-0025 15072-1000 15072-5000
TS15072-0025EA 377 USD
TS15072-0025 TS15072-1000 TS15072-5000
4-Acetamidobenzenesulphonyl chloride 99%
4-Acetamidobenzenesulphonyl chloride
Formula: C₈H₈ClNO₃S
Boiling Pt: 416 °C (1013 hPa)
Melting Pt: 145…148 °C (dec.)
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007442
CAS Number: 121-60-8
EINECS: 204-485-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to cream to light beige Crystalline or granular powder or crystals
Infrared spectrum Conforms
Melting point 144 to 148 °C (Dec)
Titration Argentometric ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR