4-Nitro-o-anisidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxy-4-nitroaniline, 2-Amino-5-nitroanisole

15064-0050 15064-1000 15064-5000
TS15064-0050EA 26 USD
TS15064-0050 TS15064-1000 TS15064-5000
4-Nitro-o-anisidine 99%
4-Nitro-o-anisidine
Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007363
CAS Number: 97-52-9
EINECS: 202-588-6

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Melting point 139 to 142 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR