Ethyl salicylate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-hydroxybenzoate
15049-0010
15049-0250
15049-2500
TS15049-0010EA
89
USD
TS15049-0010
TS15049-0050
TS15049-0250
TS15049-2500
Ethyl salicylate 99+%
Ethyl salicylate
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 234 Melting Pt: 1 Density: 1.128 g/cm³ Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002215 CAS Number: 118-61-6 EINECS: 204-265-5 Merck Index: 13,03881 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5210 to 1.5250 (20°C, 589 nm) |
GC | ≥99.0 % |
Specific gravity | (25/25°C) 1.126 to 1.13 |
Acid value | ≤1 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...