α,4-Dibromotoluene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Bromobenzyl bromide, 1-Bromo-4-(bromomethyl)benzene

15041-0250 15041-1000
TS15041-0250EA 52 USD
TS15041-0250 TS15041-1000
α,4-Dibromotoluene 98%
α,4-Dibromotoluene
Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 115…124 °C (12 mmHg)
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00000179
CAS Number: 589-15-1
EINECS: 209-636-5
UN: 1759
ADR: 8,II
Merck Index: 13,01393

Order Now

Specification Test Results

Appearance White to orange to tan Powder or crystals or needles and/or chunks
Infrared spectrum Conforms
Melting point 60 to 64 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR