1-Bromo-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

15035-0050 15035-0250 15035-1000
TS15035-0050EA 84 USD
TS15035-0050 TS15035-0250 TS15035-1000
1-Bromo-2-nitrobenzene 98%
1-Bromo-2-nitrobenzene
Formula: C₆H₄BrNO₂
Boiling Pt: 261 °C (760 mmHg)
Melting Pt: 40…43 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007045
CAS Number: 577-19-5
EINECS: 209-409-0
UN: 3459
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Yellow Fused solid or crystals and/or chunks
Infrared spectrum Conforms
Melting point 39 to 43 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR