Ethyl 4-hydroxybenzoate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate

15025-0010 15025-0051 15025-1000 15025-2500
TS15025-0010EA 151 USD
TS15025-0010 TS15025-0051 TS15025-1000 TS15025-2500
Ethyl 4-hydroxybenzoate 99%
Ethyl 4-hydroxybenzoate
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 297…298 °C (760 mmHg)
Melting Pt: 115…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS Number: 120-47-8
EINECS: 204-399-4
Merck Index: 13,03869

Order Now

Specification Test Results

Appearance White to light cream Crystalline powder
Infrared spectrum Conforms
Melting point 115 to 118 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR