4-Chlorobenzoyl chloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Chlorobenzoyl chloride

15019-0025 15019-0050 15019-1000 15019-5000
TS15019-0025EA 511 USD
TS15019-0025 TS15019-0050 TS15019-1000 TS15019-5000
4-Chlorobenzoyl chloride 99+%
4-Chlorobenzoyl chloride
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 222 °C (760 mmHg)
Melting Pt: 12…14 °C
Density: 1.37 g/cm³
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00000686
CAS Number: 122-01-0
EINECS: 204-515-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20°C, 589 nm)
GC ≥99.0 %
Titration RCOCl ≥98.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR