trans-β-Methylstyrene 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: trans-Propenylbenzene, trans-1-Phenyl-1-propene

15015-0010 15015-0100 15015-0500
TS15015-0010EA 43 USD
TS15015-0010 TS15015-0100 TS15015-0500
trans-β-Methylstyrene 97% stabilized
trans-beta-Methylstyrene
Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 175 °C (760 mmHg)
Density: 0.911 g/cm³
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009280
CAS Number: 873-66-5
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5480 to 1.5500 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.01 % BHT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR