(±)-4-Methyl-2-pentanol 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (±)-Methyl isobutyl carbinol

14938-0010 14938-0025 14938-0050 14938-0100 14938-5000
TS14938-0010EA 45 USD
TS14938-0010 TS14938-0025 TS14938-0050 TS14938-0100 TS14938-5000
(±)-4-Methyl-2-pentanol 99+%
(±)-4-Methyl-2-pentanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 132 °C (760 mmHg)
Melting Pt: –90 °C
Density: 0.8 g/cm³
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00004550
CAS Number: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4100 to 1.4120 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation +28° to +32° (20°C, 589 nm) (c=1 in DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR