1-Hydroxy-2-naphthoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Hydroxy-naphthalene-2-carboxylic acid

14923-0050 14923-1000
TS14923-0050EA 20 USD
TS14923-0050 TS14923-1000
1-Hydroxy-2-naphthoic acid 98%
1-Hydroxy-2-naphthoic acid
Formula: C₁₁H₈O₃
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 199…205 °C
Flash Pt: >150 °C
MDL Number: MFCD00003960
CAS Number: 86-48-6
EINECS: 201-674-0
Merck Index: 13,04859

Order Now

Specification Test Results

Appearance Cream to beige to grey-brown Powder
Infrared spectrum Conforms
Melting point 195 to 205 °C (Dec)
Titration with NaOH ≥97.5 %
Specific optical rotation -36.0° to -40.0° (20 °C, 589 nm) (c=1 in dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR