o-Anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Salicylaldehyde methyl ether, 2-Methoxybenzaldehyde, 2-Anisaldehyde

14922-0050 14922-1000 14922-5000
TS14922-0050EA 28 USD
TS14922-0050 TS14922-1000 TS14922-5000
o-Anisaldehyde 98%
o-Anisaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 34…40 °C
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD00003308
CAS Number: 135-02-4
EINECS: 205-171-7

Order Now

Specification Test Results

Appearance Light yellow to light brown Crystals or solid or liquids
Infrared spectrum Conforms
Melting point 34 to 40 °C
GC ≥97.5 %
Specific optical rotation +19.0° to +21.0° (20 °C, 589 nm) (c=2 in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR