2-Chlorophenylhydrazinium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
14914-0050
14914-0250
TS14914-0050EA
30
USD
TS14914-0050
TS14914-0250
2-Chlorophenylhydrazinium chloride 97%
2-Chlorophenylhydrazinium chloride
Formula:
C₆H₈Cl₂N₂ Melting Pt: 186…188 °C |
MDL Number:
MFCD00012928 CAS Number: 41052-75-9 EINECS: 255-194-1 UN: 3077 |
Specification Test Results
Appearance | White to light pink to tan Powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | ≥96.0 % |
HPLC | ≥96.0 % |
TLC | ≥96.0 % |
Specific optical rotation | -2.9° to -3.9° (20°C, 589 nm) (c=1 in DMF) |
Residual solvents | ≤15.0 % EtOAc |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...