4-Bromobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Bromobenzoic Acid

14870-0250 14870-1000 14870-5000
TS14870-0250EA 55 USD
TS14870-0250 TS14870-1000 TS14870-5000
4-Bromobenzoic acid 97%
4-Bromobenzoic acid
Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Melting Pt: 251…256 °C
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7
Merck Index: 13,01392

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 251 to 257 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR