2,4-Xylidine 99%

Supplier: Thermo Scientific Chemicals
Danger

14857-0010 14857-1000
TS14857-0010EA 152 USD
TS14857-0010 TS14857-1000
2,4-Xylidine 99%
2,4-Xylidine
Formula: C₈H₁₁N
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 16 °C
Density: 0.98 g/cm³
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00007738
CAS Number: 95-68-1
EINECS: 202-440-0
UN: 1711
ADR: 6.1,II
Merck Index: 12,10217

Order Now

Specification Test Results

Appearance Clear yellow to red-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5580 to 1.5600 (20°C, 589 nm)
GC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR