Benzenesulfonamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

14849-0025 14849-0050 14849-1000 14849-5000
TS14849-0025EA 358 USD
TS14849-0025 TS14849-0050 TS14849-1000 TS14849-5000
Benzenesulfonamide 98+%
Benzenesulphonamide
Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 151…153 °C
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00007930
CAS Number: 98-10-2
EINECS: 202-637-1

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 150 to 154 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR