α-Bromo-2-nitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α-Bromo-o-nitrotoluene, 1-(Bromomethyl)-2-nitrobenzene, alpha-Bromo-2-nitrotoluene, 2-Nitrobenzyl bromide

14831-0050 14831-0250
TS14831-0050EA 56 USD
TS14831-0050 TS14831-0250
α-Bromo-2-nitrotoluene 97%
α-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
Melting Pt: 44…48 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 44 to 48 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR