(±)-3-Chloro-1,2-propanediol 99%

Supplier: Thermo Scientific Chemicals
Danger

14812-0010 14812-0025 14812-0250 14812-5000
TS14812-0010EA 113 USD
TS14812-0010 TS14812-0025 TS14812-0250 TS14812-5000
(±)-3-Chloro-1,2-propanediol 99%
(±)-3-Chloro-1,2-propanediol
Formula: C₃H₇ClO₂
Boiling Pt: 213 °C (760 mmHg)
Melting Pt: –40 °C
Density: 1.322 g/cm³
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00004712
CAS Number: 96-24-2
EINECS: 202-492-4
UN: 2689
ADR: 6.1,III
Merck Index: 13,02164

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4790 to 1.4810 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR