2-Chloro-6-nitrobenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

14775-0010 14775-0050
TS14775-0010EA 69 USD
TS14775-0010 TS14775-0050
2-Chloro-6-nitrobenzaldehyde 98%
2-Chloro-6-nitrobenzaldehyde
Formula: C₇H₄ClNO₃
Melting Pt: 69…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00007204
CAS Number: 6361-22-4

Order Now

Specification Test Results

Appearance White to yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 68 to 73 °C
GC ≥97.5 %
NMR consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR