2,4-Dichlorophenol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,3-Dichloro-4-hydroxybenzene, 1-Hydroxy-2,4-dichlorobenzene

14772-0050 14772-1000 14772-5000
TS14772-0050EA 23 USD
TS14772-0050 TS14772-1000 TS14772-5000
2,4-Dichlorophenol 99%
2,4-Dichlorophenol
Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 41…44 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002169
CAS Number: 120-83-2
EINECS: 204-429-6
UN: 2928
ADR: 6.1,II
Merck Index: 13,03098

Order Now

Specification Test Results

Appearance White to beige Crystalline solid
Infrared spectrum Conforms
Melting point 41 to 44 °C
Water ≤0.3 %
GC ≥98.5 %
Specific optical rotation +9.0° to +11.0° (20 °C, 589 nm) (C=2 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR