Pentafluorophenol 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3,4,5,6-Pentafluorophenol

14713-0050 14713-0100 14713-1000
TS14713-0050EA 42 USD
TS14713-0050 TS14713-0100 TS14713-1000
Pentafluorophenol 99+%
Pentafluorophenol
Formula: C₆HF₅O
MW: 184.07 g/mol
Boiling Pt: 143 °C (760 mmHg)
Melting Pt: 34…36 °C
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002156
CAS Number: 771-61-9
EINECS: 212-235-8
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance White to off-white Low melting crystalline solid
Infrared spectrum Conforms
GC ≥99.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR