Ethyl-4-fluorobenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Fluorobenzoic acid ethyl ester, Ethyl 4-fluorobenzoate

14680-0050 14680-0250 14680-1000
TS14680-0050EA 17 USD
TS14680-0050 TS14680-0250 TS14680-1000
Ethyl-4-fluorobenzoate 99%
Ethyl-4-fluorobenzoate
Formula: C₉H₉FO₂
Boiling Pt: 210 °C (760 mmHg)
Melting Pt: 25…27 °C
Density: 1.14 g/cm³
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00000351
CAS Number: 451-46-7
EINECS: 207-194-8
Merck Index: 13,04198

Order Now

Specification Test Results

Appearance Colorless to light yellow Low melting solid
Infrared spectrum Conforms
Refractive index 1.4850 to 1.4870 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR