Triphenylmethane 99+%

Supplier: Thermo Scientific Chemicals

14638-0250 14638-1000 14638-5000
TS14638-0250EA 38 USD
TS14638-0250 TS14638-1000 TS14638-5000
Triphenylmethane 99+%
Triphenylmethane
Formula: C₁₉H₁₆
MW: 244.34 g/mol
Boiling Pt: 358…359 °C (754 mmHg)
Melting Pt: 90…94 °C
Density: 1.01 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00004763
CAS Number: 519-73-3
EINECS: 208-275-0
Merck Index: 12,09871

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR