3-Bromophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

14634-0100 14634-0250 14634-1000
TS14634-0100EA 47 USD
TS14634-0100 TS14634-0250 TS14634-1000
3-Bromophenol 98%
3-Bromophenol
Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 236 °C (760 mmHg)
Melting Pt: 30…32 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002253
CAS Number: 591-20-8
EINECS: 209-706-5
Merck Index: 13,01413

Order Now

Specification Test Results

Appearance Clear colorless to yellow to brown Liquid after melting or solid
Infrared spectrum Conforms
Refractive index 1.5950 to 1.6010 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR