4-Bromoaniline 99+%

Supplier: Thermo Scientific Chemicals
Danger

14628-0250 14628-1000 14628-5000
TS14628-0250EA 108 USD
TS14628-0250 TS14628-1000 TS14628-5000
4-Bromoaniline 99+%
4-Bromoaniline
Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 230…250 °C
Melting Pt: 60…64 °C
Density: 1.497 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00007822
CAS Number: 106-40-1
EINECS: 203-393-9
UN: 2811
ADR: 6.1,III
Merck Index: 13,01388

Order Now

Specification Test Results

Appearance Off-white to grey-brown Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 64 °C
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR