4,4'-Dimethoxytrityl chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DMT-Cl, 4,4'-Dimethoxytriphenylmethyl chloride

14602-0050 14602-0250 14602-1000
TS14602-0050EA 59 USD
TS14602-0050 TS14602-0250 TS14602-1000
4,4'-Dimethoxytrityl chloride 98%
4,4'-Dimethoxytrityl chloride
Formula: C₂₁H₁₉ClO₂
MW: 338.8 g/mol
Melting Pt: 121…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008409
CAS Number: 40615-36-9
EINECS: 255-002-6

Order Now

Specification Test Results

Appearance Pink Powder
Infrared spectrum Conforms
Melting point 121 to 127 °C
Titration Argentometric ≥97.5 % (ex Cl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR