3,4-Dimethoxybenzylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
14083-0050
14083-0250
14083-1000
TS14083-0050EA
51
USD
TS14083-0050
TS14083-0250
TS14083-1000
3,4-Dimethoxybenzylamine 97%
3,4-Dimethoxybenzylamine
Formula:
C₉H₁₃NO₂ Boiling Pt: 281…284 °C (760 mmHg) Density: 1.1 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008116 CAS Number: 5763-61-1 EINECS: 227-287-7 UN: 2735 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5530 to 1.5570 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...