Triphenylmethyl mercaptan 97%

Supplier: Thermo Scientific Chemicals
Warning

14041-0250 14041-1000 14041-5000
TS14041-0250EA 88 USD
TS14041-0250 TS14041-1000 TS14041-5000
Triphenylmethyl mercaptan 97%
Triphenylmethyl mercaptan
Formula: C₁₉H₁₆S
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00004854
CAS Number: 3695-77-0
EINECS: 223-020-3

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 102 to 109 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR