Triphenylene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 9,10-Benzophenanthrene, Isochrysene

14036-0010 14036-0050
TS14036-0010EA 75 USD
TS14036-0010 TS14036-0050
Triphenylene 98%
Triphenylene
Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 438 °C (1013 hPa)
Melting Pt: 196…200 °C
MDL Number: MFCD00001108
CAS Number: 217-59-4
EINECS: 205-922-9
UN: 3077
ADR: 9,III
Merck Index: 12,09870

Order Now

Specification Test Results

Appearance White to beige Crystalline needles
Infrared spectrum Conforms
Melting point 196 to 200 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR