Triphenylacetic acid 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2,2,2-Triphenylacetic acid

14030-0050 14030-0100
TS14030-0050EA 74 USD
TS14030-0050 TS14030-0100
Triphenylacetic acid 99%
Triphenylacetic acid
Formula: C₂₀H₁₆O₂
MW: 288.35 g/mol
Boiling Pt: 437 °C (1013 hPa)
Melting Pt: 268…269 °C
MDL Number: MFCD00004185
CAS Number: 595-91-5
EINECS: 209-873-4

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 267 to 275 °C
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR