2,4,6-Trimethylphenol 99%

Supplier: Thermo Scientific Chemicals
Danger

14021-0100 14021-0500
TS14021-0100EA 36 USD
TS14021-0100 TS14021-0500
2,4,6-Trimethylphenol 99%
2,4,6-Trimethylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 220 °C (760 mmHg)
Melting Pt: 70…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00002235
CAS Number: 527-60-6
EINECS: 208-419-2
UN: 2430
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow to beige Powder or crystals or needles and/or chunks
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥98.5 %
Specific optical rotation -16° to -20° (20 °C, 589 nm) c=1, DMF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR