2,3,5-Trimethylhydroquinone 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,3,5-Trimethylbenzene-1,4-diol, Trimethylhydroquinone
14015-1000
14015-5000
TS14015-1000EA
52
USD
TS14015-1000
TS14015-5000
2,3,5-Trimethylhydroquinone 97%
2,3,5-Trimethylhydroquinone
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol Boiling Pt: 295 °C (1013 hPa) Melting Pt: 169…174 °C Flash Pt: 191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002346 CAS Number: 700-13-0 EINECS: 211-838-3 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to beige to orange Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 168 to 174 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...